missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzyl bromide, 97%

Catalog No. p-4229181
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC440770050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. AC440770050 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 440770050

CAS: 53413-67-5 | C9H10BrNO4 | 276.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53413-67-5
Molecular Formula C9H10BrNO4
Molecular Weight (g/mol) 276.09
InChI Key UEKFEYNZISYRRH-UHFFFAOYSA-N
Synonym 1-bromomethyl-4,5-dimethoxy-2-nitrobenzene, 4,5-dimethoxy-2-nitrobenzyl bromide, 1-bromomethyl-2-nitro-4,5-dimethoxybenzene, bmndmb, benzene, 1-bromomethyl-4,5-dimethoxy-2-nitro, 6-nitroveratryl bromide, acmc-1ao6p, 4,5-dimenoxy-2-nitrobenzyl bromide, 2-nitro-4,5-dimethoxybenzyl bromide, 1-bromomethyl-4,5-dimethoxynitrobenzene
PubChem CID 3016812
IUPAC Name 1-(bromomethyl)-4,5-dimethoxy-2-nitrobenzene
SMILES COC1=C(C=C(C(=C1)CBr)[N+](=O)[O-])OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 130°C to 132°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 276.09
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 4, 5-Dimethoxy-2-nitrobenzyl bromide
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Danger
missing translation for 'healthHazard2' GHS H Statement:
Causes severe skin burns and eye damage.
missing translation for 'healthHazard3' GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only