missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzyl Alcohol 98.0+%, TCI America™

Catalog No. D48111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48111G 1 g
D48115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48111G Supplier TCI America Supplier No. D48111G

Chemical Identifiers

CAS 1016-58-6
Molecular Formula C9H11NO5
Molecular Weight (g/mol) 213.189
MDL Number MFCD00014701
InChI Key WBSCOJBVYHQOFB-UHFFFAOYSA-N
Synonym 4,5-dimethoxy-2-nitrobenzyl alcohol, 6-nitroveratryl alcohol, 4,5-dimethoxy-2-nitrophenyl methanol, benzenemethanol, 4,5-dimethoxy-2-nitro, 2-nitro-4,5-dimethoxybenzyl alcohol, 3,4-dimethoxy-6-nitrobenzyl alcohol, 4,5-dimethoxy-2-nitrophenyl methan-1-ol, 6-nitroveratrylalcohol, pubchem8715, 4,5-dimethoxy 2-nitrobenzyl alcohol
PubChem CID 66097
IUPAC Name (4,5-dimethoxy-2-nitrophenyl)methanol
SMILES COC1=C(C=C(C(=C1)CO)[N+](=O)[O-])OC

Specifications

Melting Point 149°C
Color Yellow
Quantity 1 g
Formula Weight 213.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dimethoxy-2-nitrobenzyl Alcohol