missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzyl alcohol, 98%

Catalog No. p-7043423
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0071906 5 g
AAL0071914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0071906 Supplier Thermo Scientific Chemicals Supplier No. L0071906

CAS: 1016-58-6 | C9H11NO5 | 213.189 g/mol

4,5-Dimethoxy-2-nitrobenzyl alcohol, is used as Reagent for the protection of alcohols, etc., as ethers which can be cleaved by photolysis involving intramolecular rearrangement to derivatives of o-nitrosobenzaldehyde, which are readily cleaved by hydrolysis. It is also used as photochemical protecting groups in organic synthesis. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dimethoxy-2-nitrobenzyl alcohol, is used as Reagent for the protection of alcohols, etc., as ethers which can be cleaved by photolysis involving intramolecular rearrangement to derivatives of o-nitrosobenzaldehyde, which are readily cleaved by hydrolysis. It is also used as photochemical protecting groups in organic synthesis. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 1016-58-6
Molecular Formula C9H11NO5
Molecular Weight (g/mol) 213.189
MDL Number MFCD00014701
InChI Key WBSCOJBVYHQOFB-UHFFFAOYSA-N
Synonym 4,5-dimethoxy-2-nitrobenzyl alcohol, 6-nitroveratryl alcohol, 4,5-dimethoxy-2-nitrophenyl methanol, benzenemethanol, 4,5-dimethoxy-2-nitro, 2-nitro-4,5-dimethoxybenzyl alcohol, 3,4-dimethoxy-6-nitrobenzyl alcohol, 4,5-dimethoxy-2-nitrophenyl methan-1-ol, 6-nitroveratrylalcohol, pubchem8715, 4,5-dimethoxy 2-nitrobenzyl alcohol
PubChem CID 66097
IUPAC Name (4,5-dimethoxy-2-nitrophenyl)methanol
SMILES COC1=C(C=C(C(=C1)CO)[N+](=O)[O-])OC

Specifications

Melting Point 144°C to 148°C
Quantity 5 g
Beilstein 1880093
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 213.19
Percent Purity 98%
Chemical Name or Material 4,5-Dimethoxy-2-nitrobenzyl alcohol
EINECSNumber 213-806-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only