missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dihydro-3(2H)-thiophenone 98.0+%, TCI America™

Catalog No. p-7127769
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35115G 5 g
D351125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35115G Supplier TCI America Supplier No. D35115G
Only null left

Chemical Identifiers

CAS 1003-04-9
Molecular Formula C4H6OS
Molecular Weight (g/mol) 102.15
MDL Number MFCD00005412
InChI Key DSXFPRKPFJRPIB-UHFFFAOYSA-N
Synonym tetrahydrothiophen-3-one, 3-thiophanone, dihydrothiophen-3 2h-one, dihydro-3 2h-thiophenone, 3-thiacyclopentanone, 3-oxotetrahydrothiophene, 3 2h-thiophenone, dihydro, 3-tetrahydrothiophenone, tetrahydrothiophene-3-one, 3-oxo-2,3,4,5-tetrahydrothiophene
PubChem CID 61252
IUPAC Name thiolan-3-one
SMILES O=C1CCSC1

Specifications

Color Yellow
Boiling Point 175°C
Quantity 5 g
Formula Weight 102.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,5-Dihydro-3(2H)-thiophenone