missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dihydro-2,5-dimethyl-4-oxofuran-3-yl Butyrate 96.0+%, TCI America™

Catalog No. D445125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D445125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D445125G Supplier TCI America Supplier No. D445125G

Chemical Identifiers

CAS 114099-96-6
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD08457803
InChI Key JOLGAERKCZYHLF-UHFFFAOYNA-N
Synonym Butyric Acid 4,5-Dihydro-2,5-dimethyl-4-oxofuran-3-yl, 4-Butyryloxy-2,5-dimethyl-3(2H)-furanone
PubChem CID 19872333
IUPAC Name 2,5-dimethyl-4-oxo-4,5-dihydrofuran-3-yl butanoate
SMILES CCCC(=O)OC1=C(C)OC(C)C1=O

Specifications

Quantity 25 g
Formula Weight 198.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,5-Dihydro-2,5-dimethyl-4-oxofuran-3-yl Butyrate