CAS RN 114099-96-6
IUPAC Name:
2,5-dimethyl-4-oxo-4,5-dihydrofuran-3-yl butanoate
Synonyms:
2,5-dimethyl-4-oxo-5H-furan-3-yl butanoate
Molecular Weight (g/mol):
198.22
Molecular Formula:
C10H14O4
InChi Key:
JOLGAERKCZYHLF-UHFFFAOYNA-N
SMILES:
CCCC(=O)OC1=C(C)OC(C)C1=O
1
–
1
of
1
results
1
4,5-Dihydro-2,5-dimethyl-4-oxofuran-3-yl Butyrate 96.0+%, TCI America™
CAS: 114099-96-6 Molecular Formula: C10H14O4 Molecular Weight (g/mol): 198.22 MDL Number: MFCD08457803 InChI Key: JOLGAERKCZYHLF-UHFFFAOYNA-N Synonym: Butyric Acid 4,5-Dihydro-2,5-dimethyl-4-oxofuran-3-yl, 4-Butyryloxy-2,5-dimethyl-3(2H)-furanone PubChem CID: 19872333 IUPAC Name: 2,5-dimethyl-4-oxo-4,5-dihydrofuran-3-yl butanoate SMILES: CCCC(=O)OC1=C(C)OC(C)C1=O