missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Difluoro-2-nitrobenzoic Acid 98.0+%, TCI America™
Supplier: TCI America D355525G
Specifications
| 4,5-Difluoro-2-nitrobenzoic Acid | |
| 165°C | |
| C7H3F2NO4 | |
| 25 g | |
| HGGRAOYTQNFGGN-UHFFFAOYSA-N | |
| 203.101 | |
| 203.10 | |
| Crystalline Powder |
| 20372-63-8 | |
| Yellow | |
| MFCD00799521 | |
| 4,5-difluoro-2-nitrobenzoic acid, 4,5-difluoro-2-nitrobenzoicacid, 3,4-difluoro-6-nitrobenzoic acid, 2-nitro-4,5-difluorobenzoic acid, 4,5-difluoro-2-nitro-benzoic acid, benzoic acid, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitrobenzenecarboxylic acid, 4,5-two fluoro-2-nitrobenzoic acid, pubchem4947, acmc-1cegm | |
| C1=C(C(=CC(=C1F)F)[N+](=O)[O-])C(=O)O | |
| 4219630 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 20372-63-8 | |
| 203.101 | |
| HGGRAOYTQNFGGN-UHFFFAOYSA-N | |
| 4219630 |
| C7H3F2NO4 | |
| MFCD00799521 | |
| 4,5-difluoro-2-nitrobenzoic acid, 4,5-difluoro-2-nitrobenzoicacid, 3,4-difluoro-6-nitrobenzoic acid, 2-nitro-4,5-difluorobenzoic acid, 4,5-difluoro-2-nitro-benzoic acid, benzoic acid, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitrobenzenecarboxylic acid, 4,5-two fluoro-2-nitrobenzoic acid, pubchem4947, acmc-1cegm | |
| C1=C(C(=CC(=C1F)F)[N+](=O)[O-])C(=O)O |
Safety and Handling
TSCA : No