missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-2-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. D35555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35555G 5 g
D355525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35555G Supplier TCI America Supplier No. D35555G

Chemical Identifiers

CAS 20372-63-8
Molecular Formula C7H3F2NO4
Molecular Weight (g/mol) 203.101
MDL Number MFCD00799521
InChI Key HGGRAOYTQNFGGN-UHFFFAOYSA-N
Synonym 4,5-difluoro-2-nitrobenzoic acid, 4,5-difluoro-2-nitrobenzoicacid, 3,4-difluoro-6-nitrobenzoic acid, 2-nitro-4,5-difluorobenzoic acid, 4,5-difluoro-2-nitro-benzoic acid, benzoic acid, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitrobenzenecarboxylic acid, 4,5-two fluoro-2-nitrobenzoic acid, pubchem4947, acmc-1cegm
PubChem CID 4219630
SMILES C1=C(C(=CC(=C1F)F)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 165°C
Color Yellow
Quantity 5 g
Formula Weight 203.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Difluoro-2-nitrobenzoic Acid