missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-1,2-phenylenediamine 97.0+%, TCI America™

Catalog No. D37291G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D37291G Supplier TCI America Supplier No. D37291G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76179-40-3
Molecular Formula C6H6F2N2
Molecular Weight (g/mol) 144.13
MDL Number MFCD00061131
InChI Key PPWRHKISAQTCCG-UHFFFAOYSA-N
Synonym 1,2-diamino-4,5-difluorobenzene, 4,5-difluoro-1,2-phenylenediamine, 4,5-difluoro-o-phenylenediamine, 2-amino-4,5-difluorophenylamine, 4,5-difluoro-benzene-1,2-diamine, 4,5-difluoro-1,2-benzenediamine, 1,2-benzenediamine,4,5-difluoro, 1,2-benzenediamine, 4,5-difluoro, pubchem4354, 2-amino-4,5-difluoroaniline
PubChem CID 2736755
IUPAC Name 4,5-difluorobenzene-1,2-diamine
SMILES NC1=CC(F)=C(F)C=C1N

Specifications

Melting Point 132°C
Quantity 1 g
Formula Weight 144.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Difluoro-1,2-phenylenediamine