missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloropyridazin-3(2H)-one, 98%

Catalog No. AAL0741306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0741306 5 g
AAL0741314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0741306 Supplier Thermo Scientific Chemicals Supplier No. L0741306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 932-22-9 | C4H2Cl2N2O | 164.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 932-22-9
Molecular Formula C4H2Cl2N2O
Molecular Weight (g/mol) 164.97
MDL Number MFCD00051504
InChI Key VJWXIRQLLGYIDI-UHFFFAOYSA-N
Synonym 4,5-dichloro-3 2h-pyridazinone, 4,5-dichloropyridazin-3 2h-one, 3 2h-pyridazinone, 4,5-dichloro, 4,5-dichloropyridazin-3-ol, 4,5-dichloro-2h-pyridazin-3-one, 4,5-dichloro-3-hydroxypyridazine, 4,5-dichloro-2,3-dihydropyridazin-3-one, 4,5-dichloro-pyridazin-3-ol, 4,5-dichloro-3-pyridazinol
PubChem CID 73247
SMILES ClC1=C(Cl)C(=O)NN=C1

Specifications

Melting Point 200°C to 204°C
Quantity 5 g
Beilstein 120959
IUPAC Name 4,5-dichloro-1H-pyridazin-6-one
Formula Weight 164.98
Percent Purity 98%
Chemical Name or Material 4,5-Dichloropyridazin-3(2H)-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UR6182000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only