missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dichloroimidazole 97.0+%, TCI America™
Supplier: TCI America D209125G
Specifications
| 4,5-Dichloroimidazole | |
| 181°C | |
| C3H2Cl2N2 | |
| 25 g | |
| QAJJXHRQPLATMK-UHFFFAOYSA-N | |
| 4,5-dichloro-1H-imidazole | |
| 140018 | |
| ≥97.0% (GC,T) |
| 15965-30-7 | |
| White-Yellow | |
| MFCD00005195 | |
| 4,5-dichloroimidazole, 1h-imidazole, 4,5-dichloro, 4,5-dichlorimidazole, pubchem12880, acmc-1bqkp, 3,4-dichloro-2h-pyrazole, ksc530c6j, 1h-imidazole,4,5-dichloro, qajjxhrqplatmk-uhfffaoysa | |
| ClC1=C(Cl)N=CN1 | |
| 136.96 | |
| 136.96 | |
| Crystalline Powder |
Chemical Identifiers
| 15965-30-7 | |
| 136.96 | |
| QAJJXHRQPLATMK-UHFFFAOYSA-N | |
| 140018 | |
| ClC1=C(Cl)N=CN1 |
| C3H2Cl2N2 | |
| MFCD00005195 | |
| 4,5-dichloroimidazole, 1h-imidazole, 4,5-dichloro, 4,5-dichlorimidazole, pubchem12880, acmc-1bqkp, 3,4-dichloro-2h-pyrazole, ksc530c6j, 1h-imidazole,4,5-dichloro, qajjxhrqplatmk-uhfffaoysa | |
| 4,5-dichloro-1H-imidazole |
Safety and Handling
TSCA : No