missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloroimidazole 97.0+%, TCI America™

Catalog No. p-7127101
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20915G 5 g
D209125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20915G Supplier TCI America Supplier No. D20915G

Chemical Identifiers

CAS 15965-30-7
Molecular Formula C3H2Cl2N2
Molecular Weight (g/mol) 136.96
MDL Number MFCD00005195
InChI Key QAJJXHRQPLATMK-UHFFFAOYSA-N
Synonym 4,5-dichloroimidazole, 1h-imidazole, 4,5-dichloro, 4,5-dichlorimidazole, pubchem12880, acmc-1bqkp, 3,4-dichloro-2h-pyrazole, ksc530c6j, 1h-imidazole,4,5-dichloro, qajjxhrqplatmk-uhfffaoysa
PubChem CID 140018
IUPAC Name 4,5-dichloro-1H-imidazole
SMILES ClC1=C(Cl)N=CN1

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 5 g
Formula Weight 136.96
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dichloroimidazole