missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dichloro-2-nitroaniline 98.0+%, TCI America™
Supplier: TCI America D404125G
Specifications
| 4,5-Dichloro-2-nitroaniline | |
| 179°C | |
| MFCD00007770 | |
| 3,4-dichloro-6-nitroaniline, benzenamine, 4,5-dichloro-2-nitro, unii-3i0g2adu1l, 2-nitro-4,5-dichloroaniline, 3i0g2adu1l, 4,5-dichloro-2-nitro-aniline, 4,5-dichloro-2-nitrobenzenamine, 4,5-dichlor-2-nitroanilin, pubchem3466, acmc-1b6pu | |
| C1=C(C(=CC(=C1Cl)Cl)[N+](=O)[O-])N | |
| 207.01 | |
| 207.01 | |
| Crystalline Powder |
| 6641-64-1 | |
| C6H4Cl2N2O2 | |
| 25 g | |
| FSGTULQLEVAYRS-UHFFFAOYSA-N | |
| 4,5-dichloro-2-nitroaniline | |
| 81149 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 6641-64-1 | |
| 207.01 | |
| FSGTULQLEVAYRS-UHFFFAOYSA-N | |
| 81149 | |
| C1=C(C(=CC(=C1Cl)Cl)[N+](=O)[O-])N |
| C6H4Cl2N2O2 | |
| MFCD00007770 | |
| 3,4-dichloro-6-nitroaniline, benzenamine, 4,5-dichloro-2-nitro, unii-3i0g2adu1l, 2-nitro-4,5-dichloroaniline, 3i0g2adu1l, 4,5-dichloro-2-nitro-aniline, 4,5-dichloro-2-nitrobenzenamine, 4,5-dichlor-2-nitroanilin, pubchem3466, acmc-1b6pu | |
| 4,5-dichloro-2-nitroaniline |
Safety and Handling
TSCA : No