missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-nitroaniline 98.0+%, TCI America™

Catalog No. p-7128069
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40415G 5 g
D404125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40415G Supplier TCI America Supplier No. D40415G
Only null left

Chemical Identifiers

CAS 6641-64-1
Molecular Formula C6H4Cl2N2O2
Molecular Weight (g/mol) 207.01
MDL Number MFCD00007770
InChI Key FSGTULQLEVAYRS-UHFFFAOYSA-N
Synonym 3,4-dichloro-6-nitroaniline, benzenamine, 4,5-dichloro-2-nitro, unii-3i0g2adu1l, 2-nitro-4,5-dichloroaniline, 3i0g2adu1l, 4,5-dichloro-2-nitro-aniline, 4,5-dichloro-2-nitrobenzenamine, 4,5-dichlor-2-nitroanilin, pubchem3466, acmc-1b6pu
PubChem CID 81149
IUPAC Name 4,5-dichloro-2-nitroaniline
SMILES C1=C(C(=CC(=C1Cl)Cl)[N+](=O)[O-])N

Specifications

Melting Point 179°C
Quantity 5 g
Formula Weight 207.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dichloro-2-nitroaniline