missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromothiophene-2-carboxaldehyde, 98%

Catalog No. p-7024682
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAA1978806 5 g
AAA1978814 25 g
2 options

Catalog No. AAA1978806

Supplier: Thermo Scientific Chemicals A1978806

Only null left

CAS: 38071-22-6 | C5H2Br2OS | 269.938 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38071-22-6
Molecular Formula C5H2Br2OS
Molecular Weight (g/mol) 269.938
MDL Number MFCD00225242
InChI Key CHRAVDVWAPGZOY-UHFFFAOYSA-N
Synonym 4,5-dibromothiophene-2-carboxaldehyde, acmc-1afpx, 4,5-dibromo-2-thenaldehyde, 4,5-dibromo-2-formylthiophene, 4,5-dibromo-2-thiophenecarbaldehyde
PubChem CID 813164
IUPAC Name 4,5-dibromothiophene-2-carbaldehyde
SMILES C1=C(SC(=C1Br)Br)C=O

Specifications

Melting Point 79°C to 81°C
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 269.94
Percent Purity 98%
Chemical Name or Material 4,5-Dibromothiophene-2-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only