CAS RN 38071-22-6
IUPAC Name:
4,5-dibromothiophene-2-carbaldehyde
Synonyms:
4,5-dibromothiophene-2-carbaldehyde
Molecular Weight (g/mol):
269.94
Molecular Formula:
C5H2Br2OS
InChi Key:
CHRAVDVWAPGZOY-UHFFFAOYSA-N
SMILES:
BrC1=C(Br)C=C(S1)C=O
1
–
1
of
1
results
1
Promotions Available
4,5-Dibromothiophene-2-carboxaldehyde, 98%
4,5-Dibromothiophene-2-carboxaldehyde, 98%
CAS: 38071-22-6 Molecular Formula: C5H2Br2OS Molecular Weight (g/mol): 269.938 MDL Number: MFCD00225242 InChI Key: CHRAVDVWAPGZOY-UHFFFAOYSA-N Synonym: 4,5-dibromothiophene-2-carboxaldehyde,acmc-1afpx,4,5-dibromo-2-thenaldehyde,4,5-dibromo-2-formylthiophene,4,5-dibromo-2-thiophenecarbaldehyde PubChem CID: 813164 IUPAC Name: 4,5-dibromothiophene-2-carbaldehyde SMILES: C1=C(SC(=C1Br)Br)C=O