missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromo-2-furoic acid, 97%

Catalog No. p-7046644
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1551706 5 g
AAL1551714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1551706 Supplier Thermo Scientific Chemicals Supplier No. L1551706
Only null left

CAS: 2434-03-9 | C5H2Br2O3 | 269.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2434-03-9
Molecular Formula C5H2Br2O3
Molecular Weight (g/mol) 269.88
MDL Number MFCD00092311
InChI Key BHUVICYZDBUMIU-UHFFFAOYSA-N
Synonym 4,5-dibromo-2-furoic acid, 2,3-dibromofuran-5-carboxylic acid, 4,5-dibromo-2-furancarboxylic acid, 2-furancarboxylicacid, 4,5-dibromo, 2-furancarboxylic acid, 4,5-dibromo, zlchem 604, acmc-209xd1, ksc201e3j, 4,5-dibromo-2-furoicacid
PubChem CID 4194832
IUPAC Name 4,5-dibromofuran-2-carboxylic acid
SMILES C1=C(OC(=C1Br)Br)C(=O)O

Specifications

Melting Point 169°C to 172°C
Quantity 5 g
Beilstein 129312
Formula Weight 269.88
Percent Purity 97%
Chemical Name or Material 4,5-Dibromo-2-furoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only