accessibility menu, dialog, popup

UserName

CAS RN 2434-03-9

IUPAC Name: 4,5-dibromofuran-2-carboxylic acid
Synonyms: 4,5-dibromofuran-2-carboxylic acid
Molecular Weight (g/mol): 269.88
Molecular Formula: C5H2Br2O3
InChi Key: BHUVICYZDBUMIU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Br)=C(Br)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results