missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromo-1,2-phenylenediamine 98.0+%, TCI America™

Catalog No. p-7128083
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40625G 5 g
D406225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40625G Supplier TCI America Supplier No. D40625G

Chemical Identifiers

CAS 49764-63-8
Molecular Formula C6H6Br2N2
Molecular Weight (g/mol) 265.94
MDL Number MFCD00795598
InChI Key TTXGKCVKGXHPRI-UHFFFAOYSA-N
Synonym 1,2-Diamino-4,5-dibromobenzene
PubChem CID 12196944
IUPAC Name 4,5-dibromobenzene-1,2-diamine
SMILES NC1=CC(Br)=C(Br)C=C1N

Specifications

Melting Point 137°C
Quantity 5 g
Formula Weight 265.94
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dibromo-1,2-phenylenediamine