missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Diaminopyrimidine 98.0+%, TCI America™
Supplier: TCI America D38581G
Specifications
| 4,5-Diaminopyrimidine | |
| 203°C | |
| C4H6N4 | |
| 1 g | |
| PPAULTVPKLVLII-UHFFFAOYSA-N | |
| pyrimidine-4,5-diamine | |
| 83703 | |
| ≥98.0% (GC,T) |
| 13754-19-3 | |
| 229°C | |
| MFCD00006113 | |
| 4,5-diaminopyrimidine, 4,5-pyrimidinediamine, pyrimidine, 4,5-diamino, 4,5-pyrimidinediamine 9ci, pubchem7057, pyrimidine,5-diamino, 4,5 diamino pyrimidine, 4,5-diamino-pyrimidine, acmc-1cfbq, 4,5-diaminopyrimidiine | |
| NC1=CN=CN=C1N | |
| 110.12 | |
| 110.12 | |
| Crystalline Powder |
Chemical Identifiers
| 13754-19-3 | |
| 110.12 | |
| PPAULTVPKLVLII-UHFFFAOYSA-N | |
| 83703 | |
| NC1=CN=CN=C1N |
| C4H6N4 | |
| MFCD00006113 | |
| 4,5-diaminopyrimidine, 4,5-pyrimidinediamine, pyrimidine, 4,5-diamino, 4,5-pyrimidinediamine 9ci, pubchem7057, pyrimidine,5-diamino, 4,5 diamino pyrimidine, 4,5-diamino-pyrimidine, acmc-1cfbq, 4,5-diaminopyrimidiine | |
| pyrimidine-4,5-diamine |
Safety and Handling
TSCA : No