accessibility menu, dialog, popup

UserName

CAS RN 13754-19-3

IUPAC Name: pyrimidine-4,5-diamine
Synonyms: 4,5-diaminopyrimidine
Molecular Weight (g/mol): 110.12
Molecular Formula: C4H6N4
InChi Key: PPAULTVPKLVLII-UHFFFAOYSA-N
SMILES: NC1=CN=CN=C1N
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)