missing translation for 'onlineSavingsMsg'
Learn More

4,5-Diaminopyrimidine, 97%

Catalog No. AC112400010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112400010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC112400010 Supplier Thermo Scientific Chemicals Supplier No. 112400010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13754-19-3 | C4H6N4 | 110.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13754-19-3
Molecular Formula C4H6N4
Molecular Weight (g/mol) 110.12
MDL Number MFCD00006113
InChI Key PPAULTVPKLVLII-UHFFFAOYSA-N
Synonym 4,5-diaminopyrimidine, 4,5-pyrimidinediamine, pyrimidine, 4,5-diamino, 4,5-pyrimidinediamine 9ci, pubchem7057, pyrimidine,5-diamino, 4,5 diamino pyrimidine, 4,5-diamino-pyrimidine, acmc-1cfbq, 4,5-diaminopyrimidiine
PubChem CID 83703
IUPAC Name pyrimidine-4,5-diamine
SMILES NC1=CN=CN=C1N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 200°C
Color Colorless
Boiling Point 229°C (32.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 24, 324
Formula Weight 110.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4, 5-Diaminopyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw
EINECSNumber 237-339-

RUO – Research Use Only