missing translation for 'onlineSavingsMsg'
Learn More

4,5-Diaminophthalonitrile 98.0+%, TCI America™

Catalog No. D4329200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4329200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4329200MG Supplier TCI America Supplier No. D4329200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 129365-93-1
Molecular Formula C8H6N4
Molecular Weight (g/mol) 158.164
MDL Number MFCD02376082
InChI Key PCKAZQYWUDIFQM-UHFFFAOYSA-N
Synonym 1,2-Diamino-4,5-dicyanobenzene
PubChem CID 676368
IUPAC Name 4,5-diaminobenzene-1,2-dicarbonitrile
SMILES C1=C(C(=CC(=C1N)N)C#N)C#N

Specifications

Melting Point 264°C
Quantity 200 mg
UN Number 3439
Formula Weight 158.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Diaminophthalonitrile