accessibility menu, dialog, popup

UserName

CAS RN 129365-93-1

IUPAC Name: 4,5-diaminobenzene-1,2-dicarbonitrile
Synonyms: 4,5-diaminobenzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 158.16
Molecular Formula: C8H6N4
InChi Key: PCKAZQYWUDIFQM-UHFFFAOYSA-N
SMILES: NC1=C(N)C=C(C#N)C(=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results