missing translation for 'onlineSavingsMsg'
Learn More

4(5)-Cyanomethylimidazole 99.0+%, TCI America™

Catalog No. C13181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13181G 1 g
C13185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13181G Supplier TCI America Supplier No. C13181G

Chemical Identifiers

CAS 18502-05-1
Molecular Formula C5H5N3
Molecular Weight (g/mol) 107.116
MDL Number MFCD00070306
InChI Key DQZBHUXBFNBJLX-UHFFFAOYSA-N
Synonym 2-1h-imidazol-4-yl acetonitrile, 1h-imidazole-5-acetonitrile, 1h-imidazole-4-acetonitrile, 2-1h-imidazol-5-yl acetonitrile, 1h-imidazol-4-yl-acetonitrile, 4 5-cyanomethylimidazole, 1h-imidazol-4-yl acetonitrile, 2-3h-imidazol-4-yl acetonitrile, 4-imidazolyl acetonitrile, 1h imidazole-4-acetonitrile
PubChem CID 571973
IUPAC Name 2-(1H-imidazol-5-yl)acetonitrile
SMILES C1=C(NC=N1)CC#N

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 107.12
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4(5)-Cyanomethylimidazole