accessibility menu, dialog, popup

UserName

NHNN

CAS RN 18502-05-1

IUPAC Name: 2-(1H-imidazol-5-yl)acetonitrile
Synonyms: 2-(3H-imidazol-4-yl)acetonitrile
Molecular Weight (g/mol): 107.12
Molecular Formula: C5H5N3
InChi Key: DQZBHUXBFNBJLX-UHFFFAOYSA-N
SMILES: N#CCC1=CN=CN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results