missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Bromopyrimidin-2-yl)morpholine, 97%

Catalog No. AC432700050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432700050 5 g Glass bottle
AC432700250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432700050 Supplier Thermo Scientific Chemicals Supplier No. 432700050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 84539-22-0 | C8H10BrN3O | 244.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84539-22-0
Molecular Formula C8H10BrN3O
Molecular Weight (g/mol) 244.09
MDL Number MFCD00483251
InChI Key CEXBOCXKAGPRHD-UHFFFAOYSA-N
Synonym 4-5-bromopyrimidin-2-yl morpholine, 5-bromo-2-morpholin-1-yl-pyrimidine, 4-5-bromo-pyrimidin-2-yl-morpholine, morpholine, 4-5-bromo-2-pyrimidinyl, 5-bromo-2-morpholinopyrimidine, 5-bromo-2-4-morpholino pyrimidine, 5-bromo-2-morpholin-4-yl pyrimidine, acmc-209pvb, 2-morpholino-5-bromopyrimidine, 5-bromo-2-morpholin-1-ylpyrimidine
PubChem CID 5144362
IUPAC Name 4-(5-bromopyrimidin-2-yl)morpholine
SMILES C1COCCN1C2=NC=C(C=N2)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 91°C to 93°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 244.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-(5-Bromopyrimidin-2-yl)morpholine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only