Learn More
4-[(5-Bromo-2-furyl)methyl]morpholine, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 71897-83-1 |
|---|---|
| Molecular Formula | C9H12BrNO2 |
| Molecular Weight (g/mol) | 246.104 |
| MDL Number | MFCD01307635 |
| InChI Key | KNWZHIWPPIXAJQ-UHFFFAOYSA-N |
| Synonym | 4-5-bromo-2-furyl methyl morpholine, 4-5-bromofuran-2-yl methyl morpholine, 4-5-bromofur-2-yl methyl morpholine, 4-5-bromo-2-furanyl-methyl morpholine, 4-5-bromo-2-furylmethyl morpholine, 4-5-bromo-furan-2-ylmethyl-morpholine, morpholine,4-5-bromo-2-furanyl methyl |
| PubChem CID | 12478687 |
| IUPAC Name | 4-[(5-bromofuran-2-yl)methyl]morpholine |
| SMILES | C1COCCN1CC2=CC=C(O2)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
CC72510DA
|
Thermo Scientific Chemicals
CC72510DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC72510DE
|
Thermo Scientific Chemicals
CC72510DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 71897-83-1 | |
| 246.104 | |
| KNWZHIWPPIXAJQ-UHFFFAOYSA-N | |
| 12478687 | |
| C1COCCN1CC2=CC=C(O2)Br |
| C9H12BrNO2 | |
| MFCD01307635 | |
| 4-5-bromo-2-furyl methyl morpholine, 4-5-bromofuran-2-yl methyl morpholine, 4-5-bromofur-2-yl methyl morpholine, 4-5-bromo-2-furanyl-methyl morpholine, 4-5-bromo-2-furylmethyl morpholine, 4-5-bromo-furan-2-ylmethyl-morpholine, morpholine,4-5-bromo-2-furanyl methyl | |
| 4-[(5-bromofuran-2-yl)methyl]morpholine |
Specifications
| 71897-83-1 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD01307635 | |
| 4-5-bromo-2-furyl methyl morpholine, 4-5-bromofuran-2-yl methyl morpholine, 4-5-bromofur-2-yl methyl morpholine, 4-5-bromo-2-furanyl-methyl morpholine, 4-5-bromo-2-furylmethyl morpholine, 4-5-bromo-furan-2-ylmethyl-morpholine, morpholine,4-5-bromo-2-furanyl methyl | |
| C1COCCN1CC2=CC=C(O2)Br | |
| 246.104 | |
| 246.1 | |
| 4-[(5-Bromo-2-furyl)methyl]morpholine |
| 96.5 | |
| 97% | |
| C9H12BrNO2 | |
| 1g | |
| KNWZHIWPPIXAJQ-UHFFFAOYSA-N | |
| 4-[(5-bromofuran-2-yl)methyl]morpholine | |
| 12478687 | |
| 97% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger