missing translation for 'onlineSavingsMsg'
Learn More

4-[(5-Bromo-2-furyl)methyl]morpholine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC72510DE

 View more versions of this product

Catalog No. CC72510DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

4-[(5-Bromo-2-furyl)methyl]morpholine
96.5
97%
C9H12BrNO2
5g
KNWZHIWPPIXAJQ-UHFFFAOYSA-N
4-[(5-bromofuran-2-yl)methyl]morpholine
12478687
97%
71897-83-1
100.0
Amber Glass Bottle
MFCD01307635
4-5-bromo-2-furyl methyl morpholine, 4-5-bromofuran-2-yl methyl morpholine, 4-5-bromofur-2-yl methyl morpholine, 4-5-bromo-2-furanyl-methyl morpholine, 4-5-bromo-2-furylmethyl morpholine, 4-5-bromo-furan-2-ylmethyl-morpholine, morpholine,4-5-bromo-2-furanyl methyl
C1COCCN1CC2=CC=C(O2)Br
246.104
246.1

Chemical Identifiers

71897-83-1
246.104
KNWZHIWPPIXAJQ-UHFFFAOYSA-N
12478687
C1COCCN1CC2=CC=C(O2)Br
C9H12BrNO2
MFCD01307635
4-5-bromo-2-furyl methyl morpholine, 4-5-bromofuran-2-yl methyl morpholine, 4-5-bromofur-2-yl methyl morpholine, 4-5-bromo-2-furanyl-methyl morpholine, 4-5-bromo-2-furylmethyl morpholine, 4-5-bromo-furan-2-ylmethyl-morpholine, morpholine,4-5-bromo-2-furanyl methyl
4-[(5-bromofuran-2-yl)methyl]morpholine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions