missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Bromo-2-fluorobenzyl)morpholine, 96%, Thermo Scientific™

$554.30 - $554.30

Chemical Identifiers

CAS 488799-67-3
Molecular Formula C11H13BrFNO
Molecular Weight (g/mol) 274.133
MDL Number MFCD20441943
InChI Key LHWHXDWETOZZBT-UHFFFAOYSA-N
Synonym 4-5-bromo-2-fluorobenzyl morpholine, 4-bromo-1-fluorophenyl-2-morphlinomethyl benzene, 4-5-bromo-2-fluorophenyl methyl morpholine, 4-5-bromo-2-fluoro-benzyl-morpholine
PubChem CID 22285325
IUPAC Name 4-[(5-bromo-2-fluorophenyl)methyl]morpholine
SMILES C1COCCN1CC2=C(C=CC(=C2)Br)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6225403
View Documents
Thermo Scientific Chemicals
H6225403
1 g
Each for $554.30
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

488799-67-3
274.133
LHWHXDWETOZZBT-UHFFFAOYSA-N
22285325
C1COCCN1CC2=C(C=CC(=C2)Br)F
C11H13BrFNO
MFCD20441943
4-5-bromo-2-fluorobenzyl morpholine, 4-bromo-1-fluorophenyl-2-morphlinomethyl benzene, 4-5-bromo-2-fluorophenyl methyl morpholine, 4-5-bromo-2-fluoro-benzyl-morpholine
4-[(5-bromo-2-fluorophenyl)methyl]morpholine

Specifications

488799-67-3
MFCD20441943
4-5-bromo-2-fluorobenzyl morpholine, 4-bromo-1-fluorophenyl-2-morphlinomethyl benzene, 4-5-bromo-2-fluorophenyl methyl morpholine, 4-5-bromo-2-fluoro-benzyl-morpholine
C1COCCN1CC2=C(C=CC(=C2)Br)F
274.133
274.13
Solid
C11H13BrFNO
1 g
LHWHXDWETOZZBT-UHFFFAOYSA-N
4-[(5-bromo-2-fluorophenyl)methyl]morpholine
22285325
96%
4-(5-Bromo-2-fluorobenzyl)morpholine
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'