missing translation for 'onlineSavingsMsg'
Learn More

4,5',8-Trimethylpsoralen, 97.5%

CAS: 3902-71-4 | C14H12O3 | 228.25 g/mol

Supplier:  Thermo Scientific Chemicals 229880010

 View more versions of this product

Catalog No. AC229880010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4, 5', 8-Trimethylpsoralen
3902-71-4
100.0
White to Beige
Authentic
Glass Bottle
MFCD00005010
15, 9907
Solubility in water: insoluble. Other solubilities: sparingly soluble in boiling water,slightly soluble in methanol,ethanol,chloroform
CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1
228.25
CHEBI:28329
97.5%
97.5%
96.5
230.0°C to 234.0°C
0.5% max. (105°C, 6 hrs)
96.5% min. (on dried substance) (HPLC)
C14H12O3
1 g
trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen
FMHHVULEAZTJMA-UHFFFAOYSA-N
2,5,9-trimethylfuro[3,2-g]chromen-7-one
5585
228.25
Crystalline Powder

Chemical Identifiers

3902-71-4
228.25
FMHHVULEAZTJMA-UHFFFAOYSA-N
5585
2,5,9-trimethylfuro[3,2-g]chromen-7-one
C14H12O3
MFCD00005010
trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen
CHEBI:28329
CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 223-459-0

RTECSNumber : LV1576000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only