missing translation for 'onlineSavingsMsg'
Learn More

4,5,6-Trifluoropyrimidine, 97%, Thermo Scientific Chemicals

Catalog No. AAH2585503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2585503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2585503 Supplier Thermo Scientific Chemicals Supplier No. H2585503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17573-78-3 | Molecular Formula: C4HF3N2 | Molecular Weight: 134.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17573-78-3
Molecular Formula C4HF3N2
Molecular Weight (g/mol) 134.06
MDL Number MFCD05662363
InChI Key AQKOJKCYBNUFLU-UHFFFAOYSA-N
Synonym pyrimidine, 4,5,6-trifluoro, pubchem6922, acmc-1bol2, pyrimidine,4,5,6-trifluoro, 4,5,6-trifluoro-1,3-diazine, 4,5,6-tris fluoranyl pyrimidine, 4,5,6-trifluoropyrimidine, 4,5,6-trifluoro pyrimidine, pyrimidine, 4,5,6-trifluoro-8ci,9ci
PubChem CID 3572764
IUPAC Name 4,5,6-trifluoropyrimidine
SMILES FC1=NC=NC(F)=C1F

Specifications

Density 1.4609
Boiling Point 83°C to 85°C
Flash Point >85°C (185°F)
Refractive Index 1.408
Quantity 1 g
UN Number UN1760
Formula Weight 134.06
Percent Purity 97%
Chemical Name or Material 4,5,6-Trifluoropyrimidine
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only