CAS RN 17573-78-3
IUPAC Name:
4,5,6-trifluoropyrimidine
Synonyms:
4,5,6-trifluoropyrimidine
Molecular Weight (g/mol):
134.06
Molecular Formula:
C4HF3N2
InChi Key:
AQKOJKCYBNUFLU-UHFFFAOYSA-N
SMILES:
FC1=NC=NC(F)=C1F
1
–
1
of
1
results
1
Promotions Available
4,5,6-Trifluoropyrimidine, 97%, Thermo Scientific Chemicals
4,5,6-Trifluoropyrimidine, 97%, Thermo Scientific Chemicals
CAS: 17573-78-3 Molecular Formula: C4HF3N2 Molecular Weight (g/mol): 134.06 MDL Number: MFCD05662363 InChI Key: AQKOJKCYBNUFLU-UHFFFAOYSA-N Synonym: pyrimidine, 4,5,6-trifluoro,pubchem6922,acmc-1bol2,pyrimidine,4,5,6-trifluoro,4,5,6-trifluoro-1,3-diazine,4,5,6-tris fluoranyl pyrimidine,4,5,6-trifluoropyrimidine,4,5,6-trifluoro pyrimidine,pyrimidine, 4,5,6-trifluoro-8ci,9ci PubChem CID: 3572764 IUPAC Name: 4,5,6-trifluoropyrimidine SMILES: FC1=NC=NC(F)=C1F