missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrothieno[3,2-c]pyridine 98.0+%, TCI America™

Catalog No. p-7137355
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26865G 5 g
T268625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26865G Supplier TCI America Supplier No. T26865G

Chemical Identifiers

CAS 54903-50-3
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.216
MDL Number MFCD03839944
InChI Key OGUWOLDNYOTRBO-UHFFFAOYSA-N
PubChem CID 3085076
IUPAC Name 4,5,6,7-tetrahydrothieno[3,2-c]pyridine
SMILES C1CNCC2=C1SC=C2

Specifications

Color Yellow
Boiling Point 75°C
Quantity 5 g
Formula Weight 139.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4,5,6,7-Tetrahydrothieno[3,2-c]pyridine
TSCA No
Recommended Storage Refrigerator