accessibility menu, dialog, popup

UserName

CAS RN 54903-50-3

IUPAC Name: 4H,5H,6H,7H-thieno[3,2-c]pyridine
Synonyms: 4H,5H,6H,7H-thieno[3,2-c]pyridine
Molecular Weight (g/mol): 139.22
Molecular Formula: C7H9NS
InChi Key: OGUWOLDNYOTRBO-UHFFFAOYSA-N
SMILES: C1CC2=C(CN1)C=CS2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results