missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrobenzo[b]thiophene-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. AAH5854803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5854803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5854803 Supplier Thermo Scientific Chemicals Supplier No. H5854803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 19156-54-8
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00652575
InChI Key TUZZQEHPGHKGRJ-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylicacid, benzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydro, benzo b thiophene-3-carboxylicacid, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic a, 4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylicacid
PubChem CID 767738
IUPAC Name 4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic acid
SMILES C1CCC2=C(C1)C(=CS2)C(=O)O

Specifications

Quantity 1 g
Beilstein 1367376
Formula Weight 182.24
Percent Purity 97%
Chemical Name or Material 4,5,6,7-Tetrahydrobenzo[b]thiophene-3-carboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures