CAS RN 19156-54-8
IUPAC Name:
4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Synonyms:
4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Molecular Weight (g/mol):
181.23
Molecular Formula:
C9H9O2S
InChi Key:
TUZZQEHPGHKGRJ-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CSC2=C1CCCC2
1
–
1
of
1
results
1
Promotions Available
4,5,6,7-Tetrahydrobenzo[b]thiophene-3-carboxylic acid, 97%, Thermo Scientific™
4,5,6,7-Tetrahydrobenzo[b]thiophene-3-carboxylic acid, 97%, Thermo Scientific™
CAS: 19156-54-8 Molecular Formula: C9H10O2S Molecular Weight (g/mol): 182.237 MDL Number: MFCD00652575 InChI Key: TUZZQEHPGHKGRJ-UHFFFAOYSA-N Synonym: 4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylic acid,4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid,4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylicacid,benzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydro,benzo b thiophene-3-carboxylicacid, 4,5,6,7-tetrahydro,4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic a,4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylicacid PubChem CID: 767738 IUPAC Name: 4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic acid SMILES: C1CCC2=C(C1)C(=CS2)C(=O)O