missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride, 95%

CAS: 128852-17-5 | C8H9ClO2S2 | 236.73 g/mol

$204.78 - $559.60

Chemical Identifiers

CAS 128852-17-5
Molecular Formula C8H9ClO2S2
Molecular Weight (g/mol) 236.73
MDL Number MFCD18825753
InChI Key UUBSHLWNADKCBU-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydrobenzo b thiophene-2-sulfonyl chloride, 2-chlorosulphonyl-4,5,6,7-tetrahydrobenzo b thiophene, 4,5,6,7-tetrahydro-1-benzothiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydrobenzo b thiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydro-1-benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonyl chloride, 4,5,6,7-tetrahydro-9ci
PubChem CID 15158650
IUPAC Name 4,5,6,7-tetrahydro-1-benzothiophene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC2=C(CCCC2)S1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH34470MD
View Documents
Thermo Scientific Chemicals
H34470MD
250 mg N/A
Only null left
 
AAH3447003
View Documents
Thermo Scientific Chemicals
H3447003
1 g N/A
Only null left
 
Description

Description

4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water

Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

128852-17-5
236.73
UUBSHLWNADKCBU-UHFFFAOYSA-N
15158650
ClS(=O)(=O)C1=CC2=C(CCCC2)S1
C8H9ClO2S2
MFCD18825753
4,5,6,7-tetrahydrobenzo b thiophene-2-sulfonyl chloride, 2-chlorosulphonyl-4,5,6,7-tetrahydrobenzo b thiophene, 4,5,6,7-tetrahydro-1-benzothiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydrobenzo b thiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydro-1-benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonyl chloride, 4,5,6,7-tetrahydro-9ci
4,5,6,7-tetrahydro-1-benzothiophene-2-sulfonyl chloride

Specifications

128852-17-5
C8H9ClO2S2
250 mg
Moisture sensitive
Slightly soluble in water
ClS(=O)(=O)C1=CC2=C(CCCC2)S1
236.73
236.74
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride
64°C to 66°C
MFCD18825753
UN3261
4,5,6,7-tetrahydrobenzo b thiophene-2-sulfonyl chloride, 2-chlorosulphonyl-4,5,6,7-tetrahydrobenzo b thiophene, 4,5,6,7-tetrahydro-1-benzothiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydrobenzo b thiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydro-1-benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonyl chloride, 4,5,6,7-tetrahydro-9ci
UUBSHLWNADKCBU-UHFFFAOYSA-N
4,5,6,7-tetrahydro-1-benzothiophene-2-sulfonyl chloride
15158650
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only