Learn More
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride, 95%
CAS: 128852-17-5 | C8H9ClO2S2 | 236.73 g/mol
Supplier: Thermo Scientific Chemicals H3447003
Description
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications
| 4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulfonyl chloride | |
| 64°C to 66°C | |
| MFCD18825753 | |
| UN3261 | |
| 4,5,6,7-tetrahydrobenzo b thiophene-2-sulfonyl chloride, 2-chlorosulphonyl-4,5,6,7-tetrahydrobenzo b thiophene, 4,5,6,7-tetrahydro-1-benzothiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydrobenzo b thiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydro-1-benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonyl chloride, 4,5,6,7-tetrahydro-9ci | |
| UUBSHLWNADKCBU-UHFFFAOYSA-N | |
| 4,5,6,7-tetrahydro-1-benzothiophene-2-sulfonyl chloride | |
| 15158650 | |
| 95% |
| 128852-17-5 | |
| C8H9ClO2S2 | |
| 1 g | |
| Moisture sensitive | |
| Slightly soluble in water | |
| ClS(=O)(=O)C1=CC2=C(CCCC2)S1 | |
| 236.73 | |
| 236.74 |
Chemical Identifiers
| 128852-17-5 | |
| 236.73 | |
| UUBSHLWNADKCBU-UHFFFAOYSA-N | |
| 15158650 | |
| ClS(=O)(=O)C1=CC2=C(CCCC2)S1 |
| C8H9ClO2S2 | |
| MFCD18825753 | |
| 4,5,6,7-tetrahydrobenzo b thiophene-2-sulfonyl chloride, 2-chlorosulphonyl-4,5,6,7-tetrahydrobenzo b thiophene, 4,5,6,7-tetrahydro-1-benzothiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydrobenzo b thiophene-2-sulphonyl chloride, 4,5,6,7-tetrahydro-1-benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonyl chloride, 4,5,6,7-tetrahydro-9ci | |
| 4,5,6,7-tetrahydro-1-benzothiophene-2-sulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only