missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrabromophenolphthalein 80.0+%, TCI America™

Catalog No. T00431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T00431G 1 g
T00435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T00431G Supplier TCI America Supplier No. T00431G

Chemical Identifiers

CAS 13027-28-6
Molecular Formula C20H10Br4O4
Molecular Weight (g/mol) 633.91
MDL Number MFCD00047314
InChI Key GNJLVMKERYSIAI-UHFFFAOYSA-N
PubChem CID 83058
IUPAC Name 4,5,6,7-tetrabromo-3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=C(Br)C(Br)=C(Br)C(Br)=C12)C1=CC=C(O)C=C1

Specifications

Quantity 1 g
Formula Weight 633.91
Percent Purity ≥80.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5,6,7-Tetrabromophenolphthalein