missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Thiomorpholinyl)aniline, 97%

Catalog No. AAH63360MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63360MD 250 mg
AAH6336003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63360MD Supplier Thermo Scientific Chemicals Supplier No. H63360MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22589-35-1 | C10H14N2S | 194.296 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22589-35-1
Molecular Formula C10H14N2S
Molecular Weight (g/mol) 194.296
MDL Number MFCD01625958
InChI Key RQXQMUUSQGCLPL-UHFFFAOYSA-N
Synonym 4-thiomorpholinoaniline, 4-thiomorpholin-4-yl aniline, 4-thiomorpholin-4-yl phenylamine, 4-thiomorpholinobenzenamine, 4-thiomorpholin-4-yl-phenylamine, benzenamine,4-4-thiomorpholinyl, benzenamine, 4-4-thiomorpholinyl, 4-p-aminophenyl-tetrahydro-4h-1,4-thiazine
PubChem CID 10774206
IUPAC Name 4-thiomorpholin-4-ylaniline
SMILES C1CSCCN1C2=CC=C(C=C2)N

Specifications

Quantity 250 mg
UN Number UN2811
Formula Weight 194.3
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(4-Thiomorpholinyl)aniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only