missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Sulfonyldiphenol, 99.7%

Catalog No. AC146915000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC146915000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC146915000 Supplier Thermo Scientific Chemicals Supplier No. 146915000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 80-09-1 | C12H10O4S | 250.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80-09-1
Molecular Formula C12H10O4S
Molecular Weight (g/mol) 250.27
MDL Number MFCD00002350
InChI Key VPWNQTHUCYMVMZ-UHFFFAOYSA-N
Synonym 4,4'-sulfonyldiphenol, bisphenol s, bis 4-hydroxyphenyl sulfone, phenol, 4,4'-sulfonylbis, 4,4'-dihydroxydiphenyl sulfone, 4-hydroxyphenyl sulfone, bis p-hydroxyphenyl sulfone, 4,4'-sulfonylbisphenol, diphone c
PubChem CID 6626
ChEBI CHEBI:34372
IUPAC Name 4-(4-hydroxyphenyl)sulfonylphenol
SMILES OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1

Specifications

CAS Min % 99.6
CAS Max % 100.0
Melting Point >245°C
Color Gray-Green to White
Infrared Spectrum Authentic
Assay Percent Range 99.6 % min. (HPLC)
Packaging Plastic bottle
Linear Formula O2S(C6H4OH)2
Quantity 500 g
Beilstein 06, 861
Solubility Information Solubility in water: 1.1g/L (20°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 250.27
Percent Purity 99.70%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Sulfonyldiphenol, 99.7%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-250-5

RUO – Research Use Only