missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Stilbenedicarboxylic acid, 96%

Catalog No. p-7081739
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1183903 1 g
AAA1183906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1183903 Supplier Thermo Scientific Chemicals Supplier No. A1183903
Only null left

CAS: 100-31-2 | C16H12O4 | 268.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-31-2
Molecular Formula C16H12O4
Molecular Weight (g/mol) 268.268
MDL Number MFCD00013994
InChI Key SBBQDUFLZGOASY-OWOJBTEDSA-N
Synonym 4,4'-stilbenedicarboxylic acid, stilbene-4,4'-dicarboxylic acid, 4-e-2-4-carboxyphenyl ethenyl benzoic acid, 4,4'-ethene-1,2-diyl dibenzoic acid, 4,4'-stilbenedicarboxylicacid, z-4,4'-ethene-1,2-diyl dibenzoic acid, 4-1e-2-4-carboxyphenyl vinyl benzoic acid, 4,4'-ethene-1,2-diyldibenzoicacid, cis-stilbene-4,4'-dicarboxylic acid, 4,4'-dicarboxystilbene
PubChem CID 5374688
IUPAC Name 4-[(E)-2-(4-carboxyphenyl)ethenyl]benzoic acid
SMILES C1=CC(=CC=C1C=CC2=CC=C(C=C2)C(=O)O)C(=O)O

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 268.27
Percent Purity 96%
Chemical Name or Material 4,4'-Stilbenedicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-838-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only