missing translation for 'onlineSavingsMsg'
Learn More

4-(4-morpholinylméthyl)benzonitrile, 97%, Thermo Scientific™

Numéro de catalogue. AC376330010
Modifier l'affichage
Click to view available options
Quantité:
1 g
Emballage:
Bouteille en verre
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC376330010 1 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC376330010 Fournisseur Thermo Scientific Chemicals Code fournisseur 376330010
Obtenez jusqu’à 1,800 $ en produits chimiques
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 37812-51-4
Formule moléculaire C12H14N2O
Poids moléculaire (g/mol) 202.26
Numéro MDL MFCD00454271
Clé InChI BCZLYNFDOJXWGN-UHFFFAOYSA-N
Synonyme 4-morpholinomethyl benzonitrile, 4-morpholin-4-ylmethyl benzonitrile, usaf a-3730, p-tolunitrile, alpha-4-morpholinyl, alpha-4-morpholinyl-p-tolunitrile, benzonitrile, 4-4-morpholinylmethyl, 4-4-morpholinylmethyl benzonitrile, 4-morpholin-4-ylmethyl benzenecarbonitrile, 4-morpholin-4-ylmethyl-benzonitrile, benzonitrile,4-4-morpholinylmethyl
PubChem CID 216864
Nom de l’IUPAC 4-(morpholine-4-ylméthyl)benzonitrile
SOURIRES C1COCCN1CC2=CC=C(C=C2)C#N

Spécifications

Nom ou substance chimique 4-(4-Morpholinylmethyl)benzonitrile
CAS % minimum 96.0
CAS % maximal 100.0
Point de fusion 84,0 °C à 89,0 °C
Couleur Du blanc au jaune
Spectre infrarouge Authentique
Fourchette de pourcentages d’essai 96% min. (GC)
Emballage Bouteille en verre
Quantité 1 g
Poids de formule 202.26
Pourcentage de pureté 97%
Forme physique Poudre cristalline
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Usage réservé à la recherche