missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Morpholinyl)aniline, 98+%

Catalog No. AAB2029206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2029206 5 g
AAB2029214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2029206 Supplier Thermo Scientific Chemicals Supplier No. B2029206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2524-67-6 | C10H14N2O | 178.235 g/mol

4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.

Solubility
Soluble in chloroform, ethyl acetate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, acid hydrides, acid chlorides, chloroformates.

Chemical Identifiers

CAS 2524-67-6
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00006169
InChI Key PHNDZBFLOPIMSM-UHFFFAOYSA-N
Synonym 4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine
PubChem CID 75655
IUPAC Name 4-morpholin-4-ylaniline
SMILES C1COCCN1C2=CC=C(C=C2)N

Specifications

Melting Point 129°C to 133°C
Quantity 5 g
UN Number UN2811
Beilstein 150957
Solubility Information Soluble in chloroform,ethyl acetate.
Formula Weight 178.24
Percent Purity ≥98%
Chemical Name or Material 4-(4-Morpholinyl)aniline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 219-760-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only