missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Methylpiperazinyl)benzoic Acid 98.0+%, TCI America™
$124.89 - $406.37
Chemical Identifiers
| CAS | 86620-62-4 |
|---|---|
| Molecular Formula | C12H16N2O2 |
| Molecular Weight (g/mol) | 220.272 |
| MDL Number | MFCD02682063 |
| InChI Key | UCFZVQHKTRSZMM-UHFFFAOYSA-N |
| Synonym | 4-4-methylpiperazin-1-yl benzoic acid, 4-4-methylpiperazino benzoic acid, 4-4-methyl-piperazin-1-yl-benzoic acid, 4-4-methylpiperazinyl benzoic acid, 4-4-methyl-piperazino benzoic acid, benzoic acid, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzoic acid, pubchem10481, ksc448o3p |
| PubChem CID | 736532 |
| IUPAC Name | 4-(4-methylpiperazin-1-yl)benzoic acid |
| SMILES | CN1CCN(CC1)C2=CC=C(C=C2)C(=O)O |
Chemical Identifiers
| 86620-62-4 | |
| 220.272 | |
| UCFZVQHKTRSZMM-UHFFFAOYSA-N | |
| 736532 | |
| CN1CCN(CC1)C2=CC=C(C=C2)C(=O)O |
| C12H16N2O2 | |
| MFCD02682063 | |
| 4-4-methylpiperazin-1-yl benzoic acid, 4-4-methylpiperazino benzoic acid, 4-4-methyl-piperazin-1-yl-benzoic acid, 4-4-methylpiperazinyl benzoic acid, 4-4-methyl-piperazino benzoic acid, benzoic acid, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzoic acid, pubchem10481, ksc448o3p | |
| 4-(4-methylpiperazin-1-yl)benzoic acid |
Specifications
| 86620-62-4 | |
| White-Yellow | |
| MFCD02682063 | |
| 4-4-methylpiperazin-1-yl benzoic acid, 4-4-methylpiperazino benzoic acid, 4-4-methyl-piperazin-1-yl-benzoic acid, 4-4-methylpiperazinyl benzoic acid, 4-4-methyl-piperazino benzoic acid, benzoic acid, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzoic acid, pubchem10481, ksc448o3p | |
| CN1CCN(CC1)C2=CC=C(C=C2)C(=O)O | |
| 220.272 | |
| 220.27 | |
| Crystalline Powder |
| 230°C | |
| C12H16N2O2 | |
| 1 g | |
| UCFZVQHKTRSZMM-UHFFFAOYSA-N | |
| 4-(4-methylpiperazin-1-yl)benzoic acid | |
| 736532 | |
| ≥98.0% (HPLC,T) | |
| 4-(4-Methylpiperazinyl)benzoic Acid |
Safety and Handling
TSCA : No