missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylpiperazinyl)benzoic Acid 98.0+%, TCI America™

Catalog No. M22671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22671G 1 g
M22675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22671G Supplier TCI America Supplier No. M22671G

Chemical Identifiers

CAS 86620-62-4
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
MDL Number MFCD02682063
InChI Key UCFZVQHKTRSZMM-UHFFFAOYSA-N
Synonym 4-4-methylpiperazin-1-yl benzoic acid, 4-4-methylpiperazino benzoic acid, 4-4-methyl-piperazin-1-yl-benzoic acid, 4-4-methylpiperazinyl benzoic acid, 4-4-methyl-piperazino benzoic acid, benzoic acid, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzoic acid, pubchem10481, ksc448o3p
PubChem CID 736532
IUPAC Name 4-(4-methylpiperazin-1-yl)benzoic acid
SMILES CN1CCN(CC1)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 1 g
Formula Weight 220.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Methylpiperazinyl)benzoic Acid