missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Methylpiperazinyl)benzaldehyde, TCI America™

$126.81 - $378.89

Chemical Identifiers

CAS 27913-99-1
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.273
InChI Key PFODEVGLOVUVHS-UHFFFAOYSA-N
Synonym 4-4-methylpiperazin-1-yl benzaldehyde, 4-4-methylpiperazino benzaldehyde, 1-4-formylphenyl-4-methylpiperazine, 4-4-methylpiperazinyl benzaldehyde, 4-4-methylpiperazino benzenecarbaldehyde, 4-4-methyl-piperazin-1-yl-benzaldehyde, benzaldehyde, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzaldehyde, pubchem20530, acmc-1cazr
PubChem CID 736533
IUPAC Name 4-(4-methylpiperazin-1-yl)benzaldehyde
SMILES CN1CCN(CC1)C2=CC=C(C=C2)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M162425G View Documents TCI America
M162425G
25g Each for $378.89
 
M16245G View Documents TCI America
M16245G
5g Each for $126.81
 
Specifications

Chemical Identifiers

27913-99-1
204.273
4-4-methylpiperazin-1-yl benzaldehyde, 4-4-methylpiperazino benzaldehyde, 1-4-formylphenyl-4-methylpiperazine, 4-4-methylpiperazinyl benzaldehyde, 4-4-methylpiperazino benzenecarbaldehyde, 4-4-methyl-piperazin-1-yl-benzaldehyde, benzaldehyde, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzaldehyde, pubchem20530, acmc-1cazr
4-(4-methylpiperazin-1-yl)benzaldehyde
C12H16N2O
PFODEVGLOVUVHS-UHFFFAOYSA-N
736533
CN1CCN(CC1)C2=CC=C(C=C2)C=O

Specifications

Undesignated
25g
C12H16N2O
PFODEVGLOVUVHS-UHFFFAOYSA-N
4-(4-methylpiperazin-1-yl)benzaldehyde
736533
Crystal-Powder at 20°C
4-(4-Methylpiperazinyl)benzaldehyde
63°C
27913-99-1
4-4-methylpiperazin-1-yl benzaldehyde, 4-4-methylpiperazino benzaldehyde, 1-4-formylphenyl-4-methylpiperazine, 4-4-methylpiperazinyl benzaldehyde, 4-4-methylpiperazino benzenecarbaldehyde, 4-4-methyl-piperazin-1-yl-benzaldehyde, benzaldehyde, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzaldehyde, pubchem20530, acmc-1cazr
CN1CCN(CC1)C2=CC=C(C=C2)C=O
204.273
204.27
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No