missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Methylpiperazinyl)benzaldehyde 98.0+%, TCI America™
Supplier: TCI America M162425G
Specifications
| 4-(4-Methylpiperazinyl)benzaldehyde | |
| 63°C | |
| MFCD00452233 | |
| 4-4-methylpiperazin-1-yl benzaldehyde, 4-4-methylpiperazino benzaldehyde, 1-4-formylphenyl-4-methylpiperazine, 4-4-methylpiperazinyl benzaldehyde, 4-4-methylpiperazino benzenecarbaldehyde, 4-4-methyl-piperazin-1-yl-benzaldehyde, benzaldehyde, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzaldehyde, pubchem20530, acmc-1cazr | |
| C[NH+]1CCN(CC1)C1=CC=C(C=O)C=C1 | |
| 205.28 | |
| 204.27 | |
| Crystalline Powder |
| 27913-99-1 | |
| C12H17N2O | |
| 25 g | |
| PFODEVGLOVUVHS-UHFFFAOYSA-O | |
| 4-(4-formylphenyl)-1-methylpiperazin-1-ium | |
| 736533 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 27913-99-1 | |
| 205.28 | |
| PFODEVGLOVUVHS-UHFFFAOYSA-O | |
| 736533 | |
| C[NH+]1CCN(CC1)C1=CC=C(C=O)C=C1 |
| C12H17N2O | |
| MFCD00452233 | |
| 4-4-methylpiperazin-1-yl benzaldehyde, 4-4-methylpiperazino benzaldehyde, 1-4-formylphenyl-4-methylpiperazine, 4-4-methylpiperazinyl benzaldehyde, 4-4-methylpiperazino benzenecarbaldehyde, 4-4-methyl-piperazin-1-yl-benzaldehyde, benzaldehyde, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzaldehyde, pubchem20530, acmc-1cazr | |
| 4-(4-formylphenyl)-1-methylpiperazin-1-ium |
Safety and Handling
TSCA : No