missing translation for 'onlineSavingsMsg'
Learn More

4'-(4-Methylphenyl)-2,2':6',2″-terpyridine, 98%

Catalog No. p-7028283
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2550977 0.1 g
AAB2550902 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2550977 Supplier Thermo Scientific Chemicals Supplier No. B2550977

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 89972-77-0 | C22H17N3 | 323.399 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89972-77-0
Molecular Formula C22H17N3
Molecular Weight (g/mol) 323.399
MDL Number MFCD06796977
InChI Key IDJYBUVHZHLIIT-UHFFFAOYSA-N
Synonym 4'-p-tolyl-2,2':6',2-terpyridine, 4'-4-methylphenyl-2,2':6',2-terpyridine, 4'-4-methylphenyl-2,2'-6',2-terpyridine, 4-4-methylphenyl-2,6-bis pyridin-2-yl pyridine, 4-4-methylphenyl-6-pyridin-2-yl-2,2'-bipyridine, 4'-4-tolyl-2,2':6',2-terpyridine, 4-p-tolyl-2,6-bis 2-pyridyl pyridine, 4-4-methylphenyl-2,6-di 2-pyridyl pyridine, 4-4-methylphenyl-2,6-dipyridin-2-ylpyridine, 2,2':6',2-terpyridine, 4'-4-methylphenyl
PubChem CID 146114
IUPAC Name 4-(4-methylphenyl)-2,6-dipyridin-2-ylpyridine
SMILES CC1=CC=C(C=C1)C2=CC(=NC(=C2)C3=CC=CC=N3)C4=CC=CC=N4

Specifications

Melting Point 167°C to 170°C
Quantity 0.1 g
Formula Weight 323.4
Percent Purity 98%
Chemical Name or Material 4'-(4-Methylphenyl)-2,2':6',2''-terpyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber YT8450000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only